missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Hexafluorphosphat, 95 %, Thermo Scientific™

Catalog Number p-7040257
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
304680-36-2
C12H23F6N2P
340.29
MFCD03427617
GRCIJNHHTXBJAK-UHFFFAOYSA-N
1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
2734243
1-Methyl-3-Octylimidazol-1-ium;Hexafluorphosphat
F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
304680-36-2
C12H23F6N2P
340.29
MFCD03427617
GRCIJNHHTXBJAK-UHFFFAOYSA-N
1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
2734243
1-Methyl-3-Octylimidazol-1-ium;Hexafluorphosphat
F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Methyl-3-n-octylimidazolium hexafluorophosphate
Menge 5 g
Empfindlichkeit Moisture sensitive
Formelmasse 340.29
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'