missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-2-methyl-4-nitroimidazol, 97 %

Catalog Number p-7027848
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
135009-57-3
C8H13N3O2
183.211
MFCD00173738
QZJVHLRSLFVNTO-UHFFFAOYSA-N
1-n-butyl-2-methyl-4-nitroimidazole, 1-butyl-2-methyl-4-nitro-1h-imidazole, 1h-imidazole,1-butyl-2-methyl-4-nitro, acmc-1bwn1, chembl95635
2801466
1-Butyl-2-Methyl-4-Nitroimidazol
CCCCN1C=C(N=C1C)[N+](=O)[O-]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
135009-57-3
C8H13N3O2
183.211
MFCD00173738
QZJVHLRSLFVNTO-UHFFFAOYSA-N
1-n-butyl-2-methyl-4-nitroimidazole, 1-butyl-2-methyl-4-nitro-1h-imidazole, 1h-imidazole,1-butyl-2-methyl-4-nitro, acmc-1bwn1, chembl95635
2801466
1-Butyl-2-Methyl-4-Nitroimidazol
CCCCN1C=C(N=C1C)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-n-Butyl-2-methyl-4-nitroimidazole
Menge 1 g
Formelmasse 183.21
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'