missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol-D7, TRC

Artikelnummer. 30477211
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30477211 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477211 Supplier Toronto Research Chemicals Supplier No. TRCN36799610MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 1-Naphthol-d7 (ring d7)
CAS 124251-84-9
Hinweise zur Reinheitsqualität HPLC
Summenformel C10 2H7 H O
Menge 10 mg
Synonym 1-Naphthalen-2,3,4,5,6,7,8-d7-ol, 1-Naphthol-d7, 1-Naphthol (ring-D7)
InChI-Formel InChI=1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H/i1D,2D,3D,4D,5D,6D,7D
SMILES [2H]c1c([2H])c([2H])c2c(O)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC-Name 2,3,4,5,6,7,8-Heptadeuterionaphthalin-1-ol
Molekulargewicht (g/mol) 151.21
Formelmasse 151.1015
Reinheit (%) >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.