missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Fos VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA1335)

Artikelnummer. 30131605
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30131605 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30131605 Lieferant Invitrogen™ Lieferanten-Nr. MA554572

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Alpaca Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

FBJ murine osteosarcoma viral oncogene homolog is commonly known as Cellular protooncogene Fos (c-Fos). It belongs to the fos transcription factor family and is encoded by the C-FOS gene located on chromosome 14. c-Fos heterodimerizes with c-Jun to form the Activator Protein -1 (AP-1) transcription factor which regulates transcription of several genes that play an important role in cellular signal transduction, cell proliferation and differentiation. c-Fos has been shown to be phosphorylated by various stimuli including growth factors and insulin at atleast seven different sites. The best known upstream kinase for phosphorylation at Threonine 232 is ERK MAPK which regulates localization of c-Fos to the nucleus and is thus important for c-Fos induced transcriptional activity.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen c-Fos VHH-8His-Cys-tag
Anwendungen ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Klassifikation Recombinant Monoclonal
Klon SAA1335
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen FOS
Gen-Zugriffsnummer P01100
Gen-Alias activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog
Gensymbole FOS
Wirtsspezies Alpaca
Immunogen Recombinant Human FOS.
Reinigungsverfahren Metal-chelate Chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 2353
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype VHH
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.