Learn More
ACROBiosystems Biotinylated Human PD-L1 / B7-H1 Protein, Fc,Avitag™,His Tag
MALS verified
Marke:  ACROBiosystems PD1-H82F3-25UG
Weitere Details : Netto-Gewicht : 0.00250kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15301179
  • 467.87 € / 25 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Programmed cell death 1 ligand 1 (PD-L1) is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses. B7-H1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. PD-L1 is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 involve in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.
This protein carries a human IgG1 Fc fragment at the C-terminus, followed by a polyhistidine tag. The Avi tag (Avitag™) is inserted in-between the Fc and his tags. The protein has a calculated MW of 54.0 kDa. The protein migrates as 66-75 kDa on a SDS-PAG
Spezifikation
Spezifikation
NP_054862.1
25 μg
Phe 19 - Arg 238
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1
Precision Avi
His Tag and Fc Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
54.0 kDa
Biotinylated Human PD-L1, Fc,Avitag,His Tag (PD1-H82F3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).
-20°C
PD-L1
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
MABSol™ Biotin Labeled Proteins