Learn More
ACROBiosystems Human IFN-alpha / beta R1 Protein, Fc Tag
Marke: ACROBiosystems IF1-H5253-100UG
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Interferon alpha/beta receptor 1 ( IFNAR1) is also known as Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1 (CRF2-1), Type I interferon receptor 1, IFNAR, which belongs to the type II cytokine receptor family. IFNAR1 /IFNAR contains four fibronectin type-III domains. IFNAR1 associates with IFNAR2 to form the type I interferon receptor. IFNAR1 is receptor for interferons alpha and beta. IFNAR1 can transduce IFNB signals without the help of IFNAR2, and not activating the Jak-STAT pathway.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000620 | |
100 μg | |
Lys 28 - Lys 436 | |
IFNAR1,IFNAR,CRF2-1,IFN-R-1,IFNAR-1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
73.8 kDa | |
Human IFNAR1, Fc Tag (IF1-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Lys 436 (Accession # NP_000620). | |
-20°C | |
IFN-alpha/beta R1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |