Learn More
ACROBiosystems Human BCMA / TNFRSF17 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Marke: ACROBiosystems BCA-H5259-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. The protein migrates as 40-47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001183.2 | |
1 mg | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
Llama IgG Fc Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
33.7 kDa | |
Human BCMA, Llama IgG2b Fc Tag, low endotoxin (BCA-H5259) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). | |
-20°C | |
BCMA | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |