Learn More
ACROBiosystems Human BTN3A3 / BTF3 Protein, His Tag
Marke: ACROBiosystems BT3-H52H0-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Butyrophilin subfamily 3 member A3 (BTN3A3),a member of the immunoglobulin superfamily and BTN/MOG family, is also known as BTF3, which contains one B30.2/SPRY domain and two Ig-like V-type (immunoglobulin-like) domains. BTN3A3 is detected in peripheral blood mononuclear cells ,T-cells ,spleen and lymphocytes. BTN3A3 also plays a role in T-cell responses in the adaptive immune response. Stimulation of human butyrophilin 3(BTN3) molecules results in negative regulation of cellular immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.5 kDa. The protein migrates as 28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH15815 | |
100 μg | |
Gln 30 - Trp 248 | |
BTN3A3,BTF3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
25.5 kDa | |
Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession # AAH15815). | |
-20°C | |
BTN3A3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |