Learn More
ACROBiosystems Mouse BTLA Protein, Fc Tag
Marke: ACROBiosystems BTA-M5253-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
42.8 kDa | |
Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession # Q32MV9). | |
-20°C | |
BTLA | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
100 μg | |
Glu 30 - Gly 176 | |
BTLA,CD272 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |