Learn More
ACROBiosystems Human CD37 Protein, Fc Tag
Marke:  ACROBiosystems CD7-H526X-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15806277
  • 383.89 € / 50 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Leukocyte antigen CD37 (CD37) is also known as Tetraspanin-26 (Tspan-26) and TSPAN26,which is a member of the transmembrane 4 superfamily or tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins encoded by CD37 gene mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD37 is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. CD37 may play a role in T-cell-B-cell interactions.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 41.6 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAI06753
50 μg
Ala 113 - Asn 240
CD37,Tspan-26,TSPAN26
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
41.6 kDa
Human CD37, Fc Tag (CD7-H526x) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Asn 240 (Accession # AAI06753).
-20°C
CD37
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens