Learn More
ACROBiosystems Human E-Cadherin / Cadherin-1 Protein, Fc Tag
Marke: ACROBiosystems ECD-H5250-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. Cadherin-1 (CDH1) is also known as epithelial cadherin (E-cadherin), CD_antigen (CD324), Uvomorulin (UVO) ECAD and CDHE, CDH1 / CD324 contains 5 cadherin domains. CDH1 / CD324 / ECAD is expressed in non-neural epithelial tissues. CDH1 / E-CAD is involved in mechanisms regulating cell-cell adhesions, mobility and proliferation of epithelial cells and has a potent invasive suppressor role. It is a ligand for integrin alpha-E/beta-7. E-Cad promotes non-amyloidogenic degradation of Abeta precursors and has a strong inhibitory effect on APP C99 and C83 production. Defects in CDH1 / CD324 / ECAD are the cause of hereditary diffuse gastric cancer (HDGC).
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.3 kDa. The protein migrates as 100 kDa and 120 kDa Cadherin, a 48 kDa Propeptide under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
AAI41839 | |
100 μg | |
Gln 23 - Pro 621 | |
Cadherin-1,CDH1,CD324,E-cadherin,Arc-1,ECAD,LCAM,UVO | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
92.3 kDa | |
Human E-Cadherin, Fc Tag (ECD-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Pro 621 (Accession # AAI41839). | |
-20°C | |
E-Cadherin | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |