Learn More
ACROBiosystems Human ENPP-2 / Autotaxin Protein, His Tag (active enzyme)
Marke: ACROBiosystems EN2-H5241-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Description
Autotaxin also known as ectonucleotide pyrophosphatase / phosphodiesterase family member 2 (E-NPP 2 or ENPP2), Extracellular lysophospholipase D (LysoPLD), Autotaxin (ATX or ATX-X), which belongs to the nucleotide pyrophosphatase / phosphodiesterase family. ENPP2 contains two SMB (somatomedin-B) domains. ENPP2 hydrolyzes lysophospholipids to produce lysophosphatidic acid (LPA) in extracellular fluids. ENPP2 can act on sphingosylphosphphorylcholine producing sphingosine-1-phosphate, a modulator of cell motility. ENPP2 stimulates migration of melanoma cells, probably via a pertussis toxin-sensitive G protein.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 94.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE).Specifications
NP_001035181 | |
50 μg | |
Asp 49 - Ile 863 | |
ENPP-2,E-NPP 2,Autotaxin,LysoPLD,ATX,PDNP2,ATX-X,NPP2,PD-IALPHA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
94.5 kDa | |
Human ENPP-2, His Tag (EN2-H5241) is expressed from human 293 cells (HEK293). It contains AA Asp 49 - Ile 863 (Accession # NP_001035181). | |
-20°C | |
ENPP-2 | |
Human | |
50 mM Tris, 150 mM NaCl, pH 7.5 | |
HEK293 | |
Enzymes |