Learn More
ACROBiosystems Human FGF R1 / CD331 Protein, His Tag
Marke: ACROBiosystems FG1-H5223-1MG
Additional Details : Netto-Gewicht : 0.00001kg
Beschreibung
Fibroblast growth factor receptor 1 (FGFR1) is also known as basic fibroblast growth factor receptor 1(BFGFR1), FMS-like tyrosine kinase, CD331, and is a receptor tyrosine kinase whose ligands are specific members of the fibroblast growth factor family. This protein is one of several fibroblast growth factor receptors, which are related proteins that are involved in important processes such as cell division, regulation of cell growth and maturation, formation of blood vessels, wound healing, and embryonic development.The FGFR1 protein spans the cell membrane, so that one end of the protein remains inside the cell and the other end projects from the outer surface of the cell. This positioning allows the FGFR1 protein to interact with specific growth factors outside the cell and to receive signals that help the cell respond to its environment. When growth factors attach to the FGFR1 protein, the receptor triggers a cascade of chemical reactions inside the cell that instruct the cell to undergo certain changes, such as maturing to take on specialized functions. The FGFR1 protein is thought to play an important role in the development of the nervous system. This protein may also help regulate the growth of long bones, such as the large bones in the arms and legs.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH18128.1 | |
1 mg | |
Arg 22 - Ile 376 | |
FGFR1,BFGFR,CD331,CEK,FGFBR,FLG,HBGFR,N-SAM,FLT2,H2,KAL2,FLJ14326 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
41.2 kDa | |
Human FGF R1, His Tag (FG1-H5223) is expressed from human 293 cells (HEK293). It contains AA Arg 22 - Ile 376 (Accession # P11362-7). | |
-20°C | |
FGF R1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |