Learn More
ACROBiosystems Mouse GPA33 / A33 Protein, His Tag
Marke: ACROBiosystems GP3-M5225-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Glycoprotein A33 (GPA33) is also known as Cell surface A33 antigen, is a single-pass type I membrane protein which is expressed in normal gastrointestinal epithelium and in 95% of colon cancers. GPA33 The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily, which contains. GPA33 may play a role in cell-cell recognition and signaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 33-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_067623.1 | |
50 μg | |
Leu 22 - Ile 235 | |
GPA33,A33 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.7 kDa | |
Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession # Q99795-1). | |
-20°C | |
GPA33 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |