missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Influenza A [A/Hong Kong/483/97 (H5N1)] Hemagglutinin (HA) Protein, His Tag

Marke:  ACROBiosystems HA1-V5229-100UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15806907

  • 335.90 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.1 kDa. The protein migrates as 66-100 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

60.1 kDa
Influenza A [A/Hong Kong/483/97 (H5N1)] HA, His Tag (HA1-V5229) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Gln 531 (Accession # AAC32099.1).
-20°C
Hemagglutinin (HA)
Influenza
PBS, pH7.4
HEK293
Viral Proteins
100 μg
Asp 17 - Gln 531
HA,Hemagglutinin
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote