Learn More
ACROBiosystems Human HSP90AA1 / HSP86 Protein, GST Tag
Marke: ACROBiosystems HS1-H513X-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Heat shock protein HSP 90-alpha (HSP90AA1 or HSP90A) is also known as Heat shock 86 kDa (HSP 86 or HSP86), Renal carcinoma antigen NY-REN-38, HSPC1, HSPCA, EL52, HSP89A, HSP90N, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP2, which belongs to the heat shock protein 90 family. HSP90AA1 undergoes a functional cycle that is linked to its ATPase activity. This cycle probably induces conformational changes in the client proteins, thereby causing their activation. HSP90AA1 interacts dynamically with various co-chaperones that modulate its substrate recognition, ATPase cycle and chaperone function.
This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 49.3 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_005339 | |
100 μg | |
Glu 535 - Asp 732 | |
HSP90AA1,HSP90A,HSP 86,HSP86,HSPC1,HSPCA,EL52,HSP89A,HSP90N,HSPCAL1,HSPCAL4,HSPN,Hsp89,Hsp90,LAP2 | |
Unconjugated | |
GST Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |
49.3 kDa | |
Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It contains AA Glu 535 - Asp 732 (Accession # NP_005339). | |
-20°C | |
HSP90AA1 | |
Human | |
50 mM HEPES, 150 mM NaCl, pH7.0 | |
E.coli | |
Bio-Markers and CD Antigens |