Learn More
ACROBiosystems Mouse LIF Protein, His Tag
Marke:  ACROBiosystems LIF-M5227-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15807277
  • 359.89 € / 50 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Leukemia inhibitory factor, or LIF, an interleukin 6 class cytokine, is a protein in cells that affects cell growth and development.Leukemia Inhibitory Factor has several functions such as cholinergic neuron differentiation, control of stem cell pluripotency, bone & fat metabolism, mitogenesis of factor dependent cell lines & promotion of megakaryocyte production in vivo. Removal of LIF pushes stem cells toward differentiation, but they retain their proliferative potential or pluripotency. Therefore LIF is used in mouse embryonic stem cell culture. It is necessary to maintain the stem cells in an undifferentiated state, however genetic manipulation of embryonic stem cells allows for LIF independent growth, notably overexpression of the gene Nanog. LIF is not required for culture of human embryonic stem cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.8 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_032527.1
50 μg
Ser 24 - Phe 203
LIF,CDF,DIA,HILDA,MLPLI
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
20.8 kDa
Mouse LIF, His Tag (LIF-M5227) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Phe 203 (Accession # P09056-1).
-20°C
LIF
Mouse
PBS, pH7.4
HEK293
Cytokines and Growth Factors