Learn More
ACROBiosystems Human PROCR / EPCR / CD201 Protein, Fc Tag
Marke: ACROBiosystems PRR-H5254-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Endothelial protein C receptor (PROCR) is also known as Activated protein C receptor (APC receptor), Endothelial cell protein C receptor, CD antigen CD201. PROCR / EPCR / CD201 is a N-glycosylated protein, with a soluble form exists. PROCR is probably released by a metalloprotease. PROCR seems to have the same activity as the membrane-bound form. PROCR / EPCR / CD201 is expressed strongly in the endothelial cells of arteries and veins in heart and lung. PROCR binds activated protein C and enhances protein C activation by the thrombin - thrombomodulin complex. PROCR plays a role in the protein C pathway controlling blood coagulation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH14451 | |
100 μg | |
Ser 18 - Thr 209 | |
PROCR,EPCR,CD201,APC receptor,CCCA,CCD41 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.6 kDa | |
Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Thr 209 (Accession # AAH14451). | |
-20°C | |
PROCR | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |