Learn More
ACROBiosystems Human SIRP gamma / CD172g Protein, Fc Tag
Marke: ACROBiosystems SIG-H5253-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Signal-regulatory protein gamma (SIRPG or SIRP-gamma) is also known as CD172 antigen-like family member B (CD172g), Signal-regulatory protein beta-2 (SIRPB2), is a single-pass type I membrane protein. SIRPG is expressed on CD4+ T-cells, CD8+ T-cells, CD56-bright natural killer (NK) cells, CD20+ cells, and all activated NK cells. SIRPG interacts with CD47. On binding with CD47, CD172g mediates cell-cell adhesion.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 67-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH64532 | |
1 mg | |
Glu 29 - Ser 364 | |
SIRPG,CD172g,SIRPB2,SIRP-gamma,SIRP-b2,SIRP-beta-2,Cripto-1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
63.7 kDa | |
Human SIRP gamma, Fc Tag (SIG-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Ser 364 (Accession # AAH64532). | |
-20°C | |
SIRP gamma | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |