missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Ebolavirus BDBV (subtype Bundibugyo, strain Uganda 2007) Small/secreted Glycoprotein (sGP) Protein, His Tag

Marke:  ACROBiosystems BEP-V5220-50UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15816047

  • 347.90 € / 50 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

EBOV encodes seven structural proteins: nucleoprotein (NP), polymerase cofactor (VP35), (VP40), GP, transcription activator (VP30), VP24, and RNA polymerase (L). GP protein contains 160-kDa envelope-attached glycoprotein (GP) and a 110 kDa secreted glycoprotein (sGP). GP is a class I fusion protein which assembles as trimers on viral surface and plays an important role in virus entry and attachment. Mature GP is a disulfide-linked heterodimer formed by two subunits, GP1 and GP2, which are generated from the proteolytical process of GP precursor (pre-GP) by cellular furin during virus assembly . GP1 is responsible for binding to the receptor(s) on target cells. Interacts with CD209/DC-SIGN and CLEC4M/DC-SIGNR which act as cofactors for virus entry into the host cell. GP2 acts as a class I viral fusion protein. GP1,2 mediates endothelial cell activation and decreases endothelial barrier function. sGP seems to possess an anti-inflammatory activity as it can reverse the barrier-decreasing effects of TNF alpha.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

34.3 kDa
Ebolavirus BDBV (subtype Bundibugyo, strain Uganda 2007) sGP, His Tag (BEP-V5220) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Arg 324 (Accession # B8XCN1).
-20°C
Glycoprotein / GP (virus)
Ebolavirus
PBS, pH7.4
HEK293
Viral Proteins
50 μg
Ile 33 - Arg 324
GP,Envelope glycoprotein,GP(1,2)
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote