Learn More
ACROBiosystems Human CD72 Protein, Fc Tag
Marke: ACROBiosystems CD2-H5251-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
B-cell differentiation antigen CD72 is also known as Lyb-2. CD72 contains one C-type lectin domain. CD72 is a protein active in the immune system of animals. CD72 consists of two identical halves, each of about 39-43 kD, and is a C-type lectin. Its primarily locus of expression is B-cells, where it appears to mediate aspects of B-cell - T-cell interaction. CD72 plays a role in B-cell proliferation and differentiation. CD72 is a ligand for CD5.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH30227 | |
100 μg | |
Arg 117 - Asp 359 | |
CD72,Lyb-2,Lyb2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
54.2 kDa | |
Human CD72, Fc Tag (CD2-H5251) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # AAH30227). | |
-20°C | |
CD72 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |