Learn More
ACROBiosystems Human ErbB3 / Her3 Protein, Strep Tag
Marke: ACROBiosystems ER3-H5288-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
ErbB3,also known as Her3 (human epidermal growth factor receptor 3), is a member of the epidermal growth factor receptor (EGFR) family of receptor tyrosine kinases. This membrane-bound glycoprotein has a neuregulin binding domain but has not an active kinase domain. It therefore can bind the ligand but cannot mediate the intracellular signal transduction through protein phosphorylation. However, it does form heterodimers with ErbB2 or other EGFR members responsible for tyrosine phosphorylation to give a receptor complex and initiate the related pathway, which lead to cell proliferation or differentiation. Overexpression of this protein has been reported in numerous cancers, including prostate, bladder, and breast tumors. This protein has different isoforms derived from alternative splicing variants, and among which, the secreted isoform lacking the intermembrane region modulates the activity of membrane-bound form.
This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 71.7 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001973.2 | |
25 μg | |
Ser 20 - Thr 643 | |
ERBB3,HER3,LCCS2,MDA-BF-1,MGC88033,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3 | |
Unconjugated | |
Strep Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
71.7 kDa | |
Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2). | |
-20°C | |
ErbB3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |