Learn More
ACROBiosystems Human FABP2 / I-FABP Protein, His Tag
Marke:  ACROBiosystems FA2-H5149-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15816697
  • 323.90 € / 200 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Fatty acid-binding protein 2 (FABP2), is also known as Fatty acid-binding protein, intestinal (FABPI), Intestinal-type fatty acid-binding protein (I-FABP). FABP2 belongs to the calycin superfamily and Fatty-acid binding protein (FABP) family. FABP2 / FABPI is expressed in the small intestine and at much lower levels in the large intestine and is highest expressed in the jejunum. FABP are thought to play a role in the intracellular transport of long-chain fatty acids and their acyl-CoA esters. FABP2 is probably involved in triglyceride-rich lipoprotein synthesis. FABP2 binds saturated long-chain fatty acids with a high affinity, but binds with a lower affinity to unsaturated long-chain fatty acids. FABP2 may also help maintain energy homeostasis by functioning as a lipid sensor.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.1 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
NP_000125.2
200 μg
Ala 2 - Asp 132
FABP2,FABPI,I-FABP
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>98% as determined by SDS-PAGE
16.1 kDa
Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession # P12104-1).
-20°C
FABP2
Human
PBS, pH7.4
E.coli
Bio-Markers and CD Antigens