Learn More
ACROBiosystems Human GAPDH / G3PDH Protein, His Tag
Marke: ACROBiosystems GAH-H5145-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is also known as Peptidyl-cysteine S-nitrosylase GAPDH, which is an enzyme that catalyzes the sixth step of glycolysis and thus serves to break down glucose for energy and carbon molecules. GAPDH has both glyceraldehyde-3-phosphate dehydrogenase and nitrosylase activities, thereby playing a role in glycolysis and nuclear functions, respectively. GAPDH can participate in nuclear events including transcription, RNA transport, DNA replication and apoptosis. Also, GAPDH can modulate the organization and assembly of the cytoskeleton. Furthermore, GAPDH is a component of the GAIT (gamma interferon-activated inhibitor of translation) complex which mediates interferon-gamma-induced transcript-selective translation inhibition in inflammation processes.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 37-38 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH01601 | |
100 μg | |
Gly 2 - Glu 335 | |
GAPDH,GAPD,CDABP0047,OK/SW-cl0.12 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
36.9 kDa | |
Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession # AAH01601). | |
-20°C | |
GAPDH | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
E.coli | |
Other Recombinant Proteins |