Learn More
ACROBiosystems Mouse EGF R Protein, His Tag
Marke: ACROBiosystems EGR-M5224-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
The epidermal growth factor receptor (EGFR; ErbB-1; HER1) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.2 kDa. The protein migrates as 86-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_997538.1 | |
1 mg | |
Leu 25 - Ser 647 | |
EGFR,ERBB,ERBB1,HER1,PIG61,mENA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
71.2 kDa | |
Mouse EGF R, His Tag (EGR-M5224) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 647 (Accession # Q01279-1). | |
-20°C | |
EGF R | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |