Learn More
ACROBiosystems Human LAP (TGF-beta 1) Protein, His Tag
Marke: ACROBiosystems LAP-H5245-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Transforming growth factor beta 1 ( TGFB1) is also known as TGF-β1, CED, DPD1, TGFB. is a polypeptide member of the transforming growth factor beta superfamily of cytokines. It is a secreted protein that performs many cellular functions, including the control of cell growth, cell proliferation, cell differentiation and apoptosis. The TGFB1 protein helps control the growth and division (proliferation) of cells, the process by which cells mature to carry out specific functions (differentiation), cell movement (motility), and the self-destruction of cells (apoptosis). The TGFB1 protein is found throughout the body and plays a role in development before birth, the formation of blood vessels, the regulation of muscle tissue and body fat development, wound healing, and immune system function. TGFB1 is particularly abundant in tissues that make up the skeleton, where it helps regulate bone growth, and in the intricate lattice that forms in the spaces between cells (the extracellular matrix). Within cells, this protein is turned off (inactive) until it receives a chemical signal to become active. TGFB1 plays an important role in controlling the immune system, and shows different activities on different types of cell, or cells at different developmental stages. Most immune cells (or leukocytes) secrete TGFB1. TGFB1 has been shown to interact with TGF beta receptor 1, LTBP1, YWHAE, EIF3I and Decorin.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 30.4 kDa. The protein migrates as 38-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000651.3 (C33S) | |
100 μg | |
Leu 30 - Arg 278 | |
LAP (TGF-beta 1),LAP (TGFB1),TGFB1,CED,DPD1,LAP,TGF-beta-1,TGFB | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
30.4 kDa | |
Human LAP (TGF-beta 1), His Tag (LAP-H5245) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (Accession # NP_000651.3 (C33S)). | |
-20°C | |
LAP (TGF-beta 1) | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |