Learn More
ACROBiosystems Human LIF R / CD118 Protein, Fc Tag
Marke: ACROBiosystems LIR-H4252-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Leukemia inhibitory factor receptor is also known as LIFR; CD118; FLJ98106; FLJ99923; LIF-R; SJS2; STWS; SWS, is the receptor for leukemia inhibitory factor (LIF). The leukemia inhibitory factor is a polyfunctional cytokine that affects the differentiation, survival, and proliferation of a wide variety of cells in the adult and the embryo. LIF action appears to be mediated through a high-affinity receptor complex composed of a low-affinity LIF binding chain (LIF receptor) and a high-affinity converter subunit, gp130. Both LIFR and gp130 are members of a family of cytokine receptors that includes components of the receptors for the majority of hematopoietic cytokines and for cytokines that affect other systems, including the ciliary neurotrophic factor, growth hormone and prolactin Defects in LIFR are the cause of Stueve-Wiedemann syndrome (SWS), a severe autosomal recessive condition and belongs to the group of the bent-bone dysplasias.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 115.5 kDa. The protein migrates as 140-190 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001121143 | |
50 μg | |
Gln 45 - Ser 833 | |
LIFR,CD118,FLJ98106,FLJ99923,SJS2,STWS,SWS | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
115.5 kDa | |
Human LIF R, Fc Tag (LIR-H4252) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - Ser 833 (Accession # P42702-1). | |
-20°C | |
LIF R | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |