Learn More
ACROBiosystems Human FGF basic Protein, Tag Free
Marke: ACROBiosystems BFF-H4117-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
FGF basic is a member of the FGF family of at least 23 related mitogenic proteins which show 35-60% amino acid conservation. FGF acidic and basic, unlike the other members of the family, lack signal peptides and are apparently secreted by mechanisms other than the classical protein secretion pathway. FGF basic has been isolated from a number of sources, including neural tissue, pituitary, adrenal cortex, corpus luteum, and placenta. This factor contains four cysteine residues, but reduced FGF basic retains full biological activity, indicating that disulfide bonds are not required for this activity. bFGF is a critical component of human embryonic stem cell culture medium; the growth factor is necessary for the cells to remain in an undifferentiated state, although the mechanisms by which it does this are poorly defined. It has been demonstrated to induce gremlin expression which in turn is known to inhibit the induction of differentiation by bone morphogenetic proteins. It is necessary in mouse-feeder cell dependent culture systems, as well as in feeder and serum-free culture systems.
This protein carries no "e;tag"e;. The protein has a calculated MW of 16.5 kDa. The protein migrates as 17 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAI66646.1 | |
1 mg | |
Pro 143 - Ser 288 | |
FGF2,BFGF,FGFB,FGF basic,HBGF-2 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
16.5 kDa | |
Human FGF basic, Tag Free (BFF-H4117) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession # P09038-4). | |
-20°C | |
FGF basic | |
Human | |
PBS, pH7.4 | |
E.coli | |
Cytokines and Growth Factors |