missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse CD47 Protein, His Tag

Marke:  ACROBiosystems CD7-M522B-100UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15836287

  • 292.55 € / 100 Mikrogramm

Verfügbar ab
02-07-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Leukocyte surface antigen CD47 is also known as Antigenic surface determinant protein OA3, Integrin-associated protein (IAP) and Protein MER6. CD47 contains 1 Ig-like V-type (immunoglobulin-like) domain. CD47 is very broadly distributed on normal adult tissues. CD47 has a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins and plays an important role in memory formation and synaptic plasticity in the hippocampus by similarity. CD47 is the receptor for SIRPA, binding to which prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. CD47 Interaction with SIRPG mediates cell-cell adhesion, enhances superantigen-dependent T-cell-mediated proliferation and costimulates T-cell activation.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 33-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

XP_006521874.1
HEK293
Mouse
15.7 kDa
100 µg
Gln 19 - Lys 140
CD47,MER6,IAP,OA3
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>90% as determined by SDS-PAGE
Mouse CD47, His Tag (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).
-20°C
CD47
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation