missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human Fc gamma RIIA / CD32a (H167) Protein, Avitag™,His Tag (SPR & BLI verified)

Biotinylated Protein

Marke:  ACROBiosystems CDA-H82E6-25UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15836347

  • 302.42 € / 25 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. The activating isoform, CD32A, is expressed on monocytes, neutrophils, platelets and dendritic cells. CD32A is expressed on many immune cell types (macrophage, neutrophil, eosinophils, platelets, dendritic cells and Langerhan cells), where inhibitory ITIM­bearing receptors may also be coexpressed and co­engaged by specific ligands. CD32A delivers an activating signal upon ligand binding, and results in the initiation of inflammatory responses including cytolysis, phagocytosis, degranulation and cytokine production. The responses can be modulated by signals from the coexpressed inhibitory receptors such as CD32B, and the strength of the signal is dependent on the ratio of expression of the activating and inhibitory receptors.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 24.0 kDa. The protein migrates as 30-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

24.0 kDa
Biotinylated Human CD32a (H167), Avitag,His Tag (SPR and BLI verified) (CDA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1 (H167)).
-20°C
Fc gamma RIIA / CD32a
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
25 μg
Ala 36 - Ile 218
CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote