Learn More
ACROBiosystems Human Cystatin D / CST5 Protein, His Tag
Marke: ACROBiosystems CS5-H5220-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Cystatin-5 (CST5) is also known as Cystatin-D, is a secreted thermostable protein, which belongs to the cystatin family. CST5 is a cysteine proteinase inhibitor that possibly plays a protective role against proteinases present in the oral cavity. The order of preference for inhibition is cathepsin S > cathepsin H > cathepsin L > cathepsin B. CST5 / cystatin-D may play a protective role against proteinases present in the oral cavity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.8 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH69514 | |
1 mg | |
Gly 21 - Val 142 | |
CST5,Cystatin-5,Cystatin-D | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
14.8 kDa | |
Human Cystatin D, His Tag (CS5-H5220) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Val 142 (Accession # AAH69514). | |
-20°C | |
Cystatin D | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |