missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IgG1 Fc Protein, Tag Free

MALS verified

Marke:  ACROBiosystems FCC-H5214-1MG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15836727

  • 803.77 € / 1 Milligramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.

This protein carries no "e;tag"e;. The protein has a calculated MW of 26.1 kDa. The protein migrates as 32-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

26.1 kDa
Human IgG Fc, Tag Free (FCC-H5214) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857-1).
-20°C
IgG1 Fc
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Other Recombinant Proteins
1 mg
Glu 99 - Lys 330
IgG1
Unconjugated
Tag Free
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote