Learn More
ACROBiosystems Human GITR / TNFRSF18 Protein, Fc Tag
Marke:  ACROBiosystems GIR-H5254-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15836847
  • 359.89 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Glucocorticoid-induced TNFR-related protein (GITR) is also known as Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), activation-inducible TNFR family receptor (AITR), CD antigen CD357, which is a member of the tumor necrosis factor receptor (TNF-R) superfamily. GITR is receptor for TNFSF18, which seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. GITR also mediated NF-kappa-B activation via the TRAF2/NIK pathway.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_004186.1
100 μg
Gln 26 - Glu 161
AITR,GITR,TNFRSF18,CD357
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
41.0 kDa
Human GITR, Fc Tag (GIR-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).
-20°C
GITR
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins