Learn More
ACROBiosystems Human LAIR2 / CD306 Protein, His Tag
Marke: ACROBiosystems LA2-H5220-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Leukocyte-associated immunoglobulin-like receptor 2 (LAIR2) is also known as CD antigen CD306. LAIR2 contains one Ig-like C2-type (immunoglobulin-like) domain. LAIR2 is a member of the immunoglobulin superfamily and was identified by its similarity to LAIR1, an inhibitory receptor present on mononuclear leukocytes. LAIR2 / CD306 is thought to be secreted and may help modulate mucosal tolerance.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 18-20 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH69366 | |
100 μg | |
Gln 22 - Pro 152 | |
LAIR2,CD306 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
14.9 kDa | |
Human LAIR-2, His Tag (LA2-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Pro 152 (Accession # AAH69366). | |
-20°C | |
LAIR-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |