Learn More
ACROBiosystems Human Lumican Protein, His Tag
Marke:  ACROBiosystems LUM-H5227-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15837287
  • 311.92 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Lumican (LUM) is also known as Keratan sulfate proteoglycan lumican, KSPG lumican,LDC and SLRR2D, which belongs to the small leucine-rich proteoglycan (SLRP) family and SLRP class II subfamily. LUM /Lumican is present in the extracellular matrix of human articular cartilage at all ages, although its abundance is far greater in the adult. In the adult cartilage LUM /Lumican exists predominantly in a glycoprotein form lacking keratan sulfate, whereas the juvenile form of the molecule is a proteoglycan. LUM participates in the maintenance of tissue homeostasis and modulates cellular functions including cell proliferation, migration, and differentiation. The expression of LUM has been correlated to the growth and metastasis of various malignancies.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_002336
100 μg
Gln 19 - Asn 338
LUM,Lumican,LDC,SLRR2D,KSPG lumican
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by reduced SDS-PAGE
37.5 kDa
Human Lumican, His Tag (LUM-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 338 (Accession # NP_002336).
-20°C
Lumican
Human
PBS, pH7.4
HEK293
Other Recombinant Proteins