Learn More
ACROBiosystems Mouse PD-L2 / B7-DC Protein, Fc Tag
Marke: ACROBiosystems PD2-M5254-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Programmed cell death 1 ligand 2 (PD-L2 or PDCD1 ligand 2) is also known as Butyrophilin B7-DC, CD antigen CD273, which belongs to the immunoglobulin superfamily or BTN/MOG family. The expression of PD-L2 is up-regulated by IFNG/IFN-gamma stimulation in monocytes and induced on dendritic cells grown from peripheral blood mononuclear cells with CSF2 and IL-4. PD-L2 Involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner. PD-L2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. As a result of glycosylation, the protein migrates as 60-70 kDa under reducing (R) condition, and 120-140 kDa under non-reducing (NR) condition (SDS-PSpezifikation
NP_067371 | |
100 μg | |
Leu 20 - Arg 219 | |
PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
49.2 kDa | |
Mouse PD-L2, Fc Tag (PD2-M5254) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371). | |
-20°C | |
PD-L2 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |