Learn More
ACROBiosystems Human Nectin-1 / PVRL1 / CD111 Protein, His Tag
Marke: ACROBiosystems PV1-H5223-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Poliovirus receptor-related protein 1 (PVRL1) is also known as Herpes virus entry mediator C (HveC), Herpesvirus Ig-like receptor (HIgR), Nectin-1, CD antigen CD111, PVRL1 / Nectin-1 / CD111 belongs to the nectin family. PVRL1 / Nectin-1 is a membrane protein with three extracellular immunoglobulin domains, a single transmembrane helix and a cytoplasmic tail. The protein can mediate Ca2+-independent cellular adhesion further characterizing it as IgSF cell adhesion molecule (IgSF CAM). PVRL1 is adhesion molecule found in a wide range of tissues. PVRL1 / Nectin-1 promotes cell-cell contacts by forming homophilic or heterophilic trans-dimers. Heterophilic interactions have been detected between PVRL1/nectin-1 and PVRL3/nectin-3 and between PVRL1/nectin-1 and PVRL4/nectin-4.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.0 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002846 | |
100 μg | |
Gln 31 - Thr 334 | |
PVRL1,Nectin-1,CD111,CLPED1,ED4,HIgR,HVEC,OFC7,PRR,PRR1,PVRR,PVRR1,SK-12 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
35.0 kDa | |
Human Nectin-1, His Tag (PV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession # NP_002846). | |
-20°C | |
Nectin-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |