Learn More
ACROBiosystems Human TRAIL R4 / TNFRSF10D Protein, His Tag
Marke: ACROBiosystems TR4-H52H5-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 10D (TNFRSF10D) is also known as Decoy receptor 2 (DcR2), TNF-related apoptosis-inducing ligand receptor 4 (TRAIL receptor 4 or TRAIL-R4), CD264,which is the single-pass type I membrane protein. TNFRSF10D is receptor for the cytotoxic ligand TRAIL, which contains a truncated death domain and hence is not capable of inducing apoptosis but protects against TRAIL-mediated apoptosis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.7 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003831 | |
100 μg | |
Ala 56 - His 211 | |
TNFRSF10D,CD264,TRAILR4,DCR2,TRUNDD | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
18.7 kDa | |
Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession # NP_003831). | |
-20°C | |
TRAIL R4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |