Learn More
ACROBiosystems Human TSLP R Protein, Fc Tag
Marke:  ACROBiosystems TSR-H525A-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15837877
  • 443.87 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Receptor for thymic stromal lymphopoietin (TSLP). CRLF2 / TSLPR is a member of the type I cytokine receptor family. Together with the interleukin 7 receptor (IL7R) and TSLP activate STAT3, STAT5, and JAK2 pathways, which control processes such as cell proliferation and development of the hematopoietic system.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 62-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_071431.2
100 μg
Gly 25 - Lys 231
TSLP R,CRLF2,IL-XR,ILXR,p2RY8/CRLF2 fusion,TSLP receptor,TSLPR,CRL2,CRLF2Y,Cytokine receptor-like 2
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
50.4 kDa
Human TSLP R, Fc Tag (TSR-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Lys 231 (Accession # Q9HC73-1).
-20°C
TSLP R
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Other Recombinant Proteins