Learn More
ACROBiosystems Human VSIG8 Protein, Fc tag
Marke: ACROBiosystems VS8-H5250-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
V-set and immunoglobulin domain containing 8 (VSIG8), also known as C1orf204, is a type I transmembrane protein of the B7 family within the Ig superfamily. VSIG8 was identified from proteomic analysis of human hair shafts. It is expressed in the hair follicle and shaft, superficial layers of the nail matrix, and superficial layers of oral epithelium.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.7 kDa. As a result of glycosylation, the protein migrates as 55-65 kDa under reducing (R) condition, and 110-130 kDa under non-reducing (NR) condition (SDS-PSpezifikation
NP_001013683.1 | |
100 μg | |
Val 22 - Gly 263 | |
VSIG8,C1orf204 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
53.7 kDa | |
Human VSIG8, Fc Tag (VS8-H5250) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1). | |
-20°C | |
VSIG8 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |