Learn More
ACROBiosystems Human RAGE / AGER Protein, Fc Tag
Marke: ACROBiosystems AGR-H5253-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Advanced glycosylation end product-specific receptor (AGER) is also known as RAGE, and is a single-pass type I membrane protein which contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. AGER is highly expressed in endothelial cells. AGER mediates interactions of advanced glycosylation end products (AGE), and also accumulates in vascular tissue in aging and at an accelerated rate in diabetes, as well as acts as a mediator of both acute and chronic vascular inflammation in conditions such as atherosclerosis and in particular as a complication of diabetes. AGER / RAGE signaling plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.8 kDa. The protein migrates as 80 - 90 kDa and 35 kDa Fc fragment under reducing (R) condition (SDS-PAGE) due to glycosylation and auto-cleavage.Spezifikation
NP_001127 | |
100 µg | |
Ala 23 - Ala 344 | |
AGER,RAGE | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>60% as determined by SDS-PAGE |
60.8 kDa | |
Human RAGE, Fc Tag (AGR-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Ala 344 (Accession # NP_001127). | |
-20°C | |
RAGE | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |