Learn More
ACROBiosystems Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag
Marke: ACROBiosystems AN7-H5252-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Angiopoietin-like 7 (ANGPTL7) is also known as Corneal-Derived Transcript 6 (CDT6), and is a secreted glycoprotein that is structurally related to the angiopoietins. It is expressed in the corneal stroma, trabecular meshwork, and sclera and is elevated in glaucoma aqueous humor. Recently, ANGPTL proteins, including ANGPTL-2, -3, -4, -5 and -7, are identified as growth factors in combination with SCF, Thrombopoietin, IGF-II and FGF acidic that enhance the expansion and engraftment of human and mouse hematopoietic stem cells.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 64.1 kDa. The protein migrates as 60 kDa and 76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_066969.1 | |
100 μg | |
Gln 27 - Pro 346 | |
ANGPTL7,CDT6 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
64.1 kDa | |
Human ANGPTL7, Mouse IgG2a Fc Tag (AN7-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1). | |
-20°C | |
ANGPTL7 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |