Learn More
ACROBiosystems Human Arginase 1 / ARG1 Protein, His Tag
Marke:  ACROBiosystems AR1-H5228-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15845927
  • 419.88 € / 50 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Arginase-1 (ARG1) is also known as Liver-type arginase, Type I arginase, which belongs to the arginase family. Arginase-1 / ARG1 is a manganese-containing enzyme. The reaction catalyzed by this enzyme is: arginine + H2O → ornithine + urea. It is the final enzyme of the urea cycle. Defects in Arginase-1 / ARG1 are the cause of argininemia (ARGIN).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.8 kDa. The protein migrates as 35-38 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
NP_000036
50 μg
Met 1 - Lys 322
ARG1,Arginase-1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
35.8 kDa
Human Arginase 1, His Tag (AR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 322 (Accession # NP_000036).
-20°C
Arginase 1
Human
50 mM Tris, 150 mM NaCl, pH7.5
HEK293
Other Recombinant Proteins