Learn More
ACROBiosystems Human CD48 / SLAMF2 Protein, His Tag
Marke: ACROBiosystems BC1-H5226-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
CD48 antigen is also known as Signaling lymphocytic activation molecule 2, SLAM family member 2 (SLAMF2), Leukocyte antigen MEM-102, BCM1 surface antigen, B-lymphocyte activation marker BLAST-1. CD48 is a 65 kDa GPI-linked protein which belongs to the CD2 subfamily of immunoglobulin superfamily molecules. CD48 antigen contains two Ig-like C2-type (immunoglobulin-like) domains and two Ig-like V-type (immunoglobulin-like) domain. CD48 interacts with CD244 in an heterophilic manner. CD48 is ligand for CD2. CD48 might facilitate interaction between activated lymphocytes. CD48 is probably involved in regulating T-cell activation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH16182 | |
100 μg | |
Gln 27 - Ser 220 | |
CD48,BCM1,SLAMF2,BLAST,BLAST1,MEM-102,TCT0.1,BCM-1,SLAMF-2,BLAST-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
23.2 kDa | |
Human CD48, His Tag (BC1-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # AAH16182). | |
-20°C | |
CD48 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |