Learn More
ACROBiosystems Biotinylated Human Siglec-3 / CD33 Protein, Avitag™,His Tag
MALS verified
Marke: ACROBiosystems CD3-H82E7-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Myeloid cell surface antigen CD33 is also known as SIGLEC3, Siglecs (sialic acid binding Iglike lectins) and GP67, is a single-pass type I membrane protein which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Human CD33 / Siglec-3 cDNA encodes a 364 amino acid (aa) polypeptide with a hydrophobic signal peptide, an N-terminal Ig-like V-type domain, one Ig-like C2-type domains, a transmembrane region and a cytoplasmic tail. CD33 / Siglec-3 usually considered myeloid-specific, but it can also be found on some lymphoid cells. In the immune response, CD33 / Siglec-3 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. CD33 / Siglec-3 induces apoptosis in acute myeloid leukemia.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 29.4 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH28152.1 | |
25 μg | |
Asp 18 - His 259 | |
CD33,SIGLEC3,gp67 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |
29.4 kDa | |
Biotinylated Human Siglec-3, Avitag,His Tag (CD3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). | |
-20°C | |
Siglec-3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |