Learn More
ACROBiosystems Biotinylated Feline FcRn / FCGRT&B2M Heterodimer Protein, His, Avitag™
Biotinylated SPR verified Protein
Marke: ACROBiosystems FCN-F82W3-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
FCGRT & B2M heterodimer protein (FcRn complex) consist of two subunits: p51 (equivalent to FCGRT), and p14 (equivalent to beta-2-microglobulin), and forms an MHC class I-like heterodimer. Fc fragment of IgG, receptor, transporter, alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.
Biotinylated Feline FCGRT&B2M Heterodimer Protein, His,Avitag (SPR verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 34.0 kDa (FCGRT) and 11.5 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus, followSpezifikation
XP_006941057.2 (FCGRT) and NP_001009876.1 (B2M) | |
200 μg | |
Ala 24 - Ser 297 (FCGRT) and Val 21 - Met 118 (B2M) | |
FcRn,FCGRT and B2M | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
34.0 kDa (FCGRT) and 11.5 kDa (B2M) | |
Biotinylated Feline FCGRT&B2M; Heterodimer Protein, His,Avitag (SPR verified) (FCN-F82W3) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) and Val 21 - Met 118 (B2M) (Accession # W5VRL5-1 (FCGRT) and Q5MGS7-1 (B2M)). | |
-20°C | |
FcRn (FCGRT and B2M) | |
Feline | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |