Learn More
ACROBiosystems Human ICAM-1 / CD54 Protein, Fc Tag
Marke: ACROBiosystems IC1-H5250-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Inter-Cellular Adhesion Molecule 1 (ICAM-1) is also known as Cluster of Differentiation 54 (CD54), is a member of the immunoglobulin superfamily, and is a cell surface glycoprotein which is typically expressed in low concentrations on endothelial cells and cells of the immune system. The protein encoded by this gene is a type of intercellular adhesion molecule continuously present in low concentrations in the membranes of leukocytes and endothelial cells. Upon cytokine stimulation, the concentrations greatly increase. ICAM-1 can be induced by interleukin-1 (IL-1) and tumor necrosis factor alpha (TNFα) and is expressed by the vascular endothelium, macrophages, and lymphocytes. ICAM-1 is a ligand for LFA-1 (integrin), a receptor found on leukocytes. When activated, leukocytes bind to endothelial cells via ICAM-1/LFA-1 and then transmigrate into tissues. ICAM-1 has been implicated in subarachnoid hemorrhage (SAH). Levels of ICAM-1 are shown to be significantly elevated in patients with SAH over control subjects in many studies. ICAM-1 expressed by respiratory epithelial cells is also the binding site for rhinovirus, the causative agent of most common colds.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 75.7 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH15969 | |
1 mg | |
Gln 28 - Glu 480 | |
ICAM1,BB2,CD54,P3.58 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |
75.7 kDa | |
Human ICAM-1, Fc Tag (IC1-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession # AAH15969). | |
-20°C | |
ICAM-1 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |