missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human IgG1 Fc Protein, Avitag™,His Tag

Biotinylated Protein

Marke:  ACROBiosystems IG1-H82E2-25UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15846987

  • 353.08 € / 25 Mikrogramm

Voraussichtliches Versanddatum: 27-08-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 28.8 kDa. As a result of glycosylation, the protein migrates as 36 kDa under reducing (R) condition, and 60-66 kDa under non-r
Spezifikation

Spezifikation

Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
MABSol™ Biotinylated Human IgG1 Fc, Avitag,His Tag (IG1-H82E2) is expressed from human HEK293 cells. It contains AA Pro 100 - Lys 330 (Accession # P01857-1 (C103S)).
-20°C
IgG1 Fc
His Tag and Avi Tag
HEK293
Human
28.8 kDa
25 µg
Pro 100 - Lys 330
IgG1
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate