Learn More
ACROBiosystems Human IgG3 Fc Protein, Tag Free
SPR verified
Marke: ACROBiosystems IG3-H5200-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Immunoglobulin G3 (IgG3) is a member of many immunoglobulin G developed and secreted by effective B cells. In wake of cutting by pepsin, IgG is divided into two F(ab)s with one antigen binding site and a high conserved Fc segment. The Fc segment bears a highly conserved N-glycosylation site. Ig gamma-3 chain C region (IgG3-Fc / IGHG3) contains two constant regions of IgG3 H chain (CH2, CH3).
This protein carries no "e;tag"e;. The protein has a calculated MW of 31.2 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
31.2 kDa | |
Human IgG3 Fc, Tag Free (SPR verified) (IG3-H5200) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 377 (Accession # P01860). | |
-20°C | |
IgG3 Fc | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
1 mg | |
Glu 99 - Lys 377 | |
Ig gamma-3 chain C region,IgG,IgG3 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |