Learn More
ACROBiosystems ActiveMax™ Human IL-6 Protein, Tag Free
Marke: ACROBiosystems IL6-H4218-20UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin 6 (IL-6) is also known as HGF, BSF2,HSF, IFNB2 and IL-6, originally identified as a B cell differentiation factor, is a multifunctional cytokine that regulates immune responses, hematopoiesis, acute phase responses, and inflammatory reactions.It is secreted by T cells, macrophages , monocytes, fibroblasts,endothelial cells,et.al. to stimulate immune response to trauma, especially burns or other tissue damage leading to inflammation. Interleukin 6 has been shown to interact with interleukin-6 receptor and glycoprotein. IL-6 is relevant to many disease processes such as diabetes,atherosclerosis, depression,Alzheimer's Disease,systemic,lupus erythematosus,prostate cancer and rheumatoid arthritis. Advanced/metastatic cancer patients have higher levels of IL-6 in their blood.Hence there is an interest in developing anti-IL-6 agents as therapy against many of these diseases.
This protein carries no "e;tag"e;. The protein has a calculated MW of 20.8 kDa. The protein migrates as 23-30 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
NP_000591 | |
20 μg | |
Val 30 - Met 212 | |
IL6,Interleukin-6,BSF2,HSF,IFNB2 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
20.8 kDa | |
ActiveMax™ Human IL-6, Tag Free (IL6-H4218) is expressed from human 293 cells (HEK293). It contains AA Val 30 - Met 212 (Accession # NP_000591). | |
-20°C | |
IL-6 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |